ENA183889231 Enamine

3',4'-dichloro-3-fluoro-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD09042237
  • Cas Number : 926235-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40761
Availability details for
Quantity
  • Availability
  • Add to Cart