ENA183889236 Enamine

4-fluoro-N-{3-[(1E)-N-hydroxyethanimidoyl]phenyl}benzamide

  • MDL number MFCD09939173
  • Cas Number : 1016876-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40769

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40769
Availability details for
Quantity
  • Availability
  • Add to Cart