ENA183889243 Enamine

N-(4-hydroxyphenyl)-5-methylthiophene-2-sulfonamide

  • MDL number MFCD11171855
  • Cas Number : 1042647-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40808
Availability details for
Quantity
  • Availability
  • Add to Cart