ENA183889255 Enamine

pentanethioamide

  • MDL number MFCD09803501
  • Cas Number : 16536-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40824
Availability details for
Quantity
  • Availability
  • Add to Cart