ENA183889268 Enamine

2-(3,5-dimethyl-1H-pyrazol-1-yl)-5-(trifluoromethyl)aniline

  • MDL number MFCD08699434
  • Cas Number : 60418-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40847
Availability details for
Quantity
  • Availability
  • Add to Cart