ENA183889279 Enamine

2-{[(4-fluorophenyl)methyl]sulfanyl}pyridine-3-carboxylic acid

  • MDL number MFCD03123099
  • Cas Number : 460736-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40871
Availability details for
Quantity
  • Availability
  • Add to Cart