ENA183889292 Enamine

3-(4-formylphenoxy)propanoic acid

  • MDL number MFCD10009898
  • Cas Number : 70170-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40890

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40890
Availability details for
Quantity
  • Availability
  • Add to Cart