ENA183889295 Enamine

N-(3-amino-4-fluorophenyl)-2-(2-oxo-1,2-dihydropyrimidin-1-yl)acetamide

  • MDL number MFCD09733207
  • Cas Number : 953732-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40894
Availability details for
Quantity
  • Availability
  • Add to Cart