ENA183889296 Enamine

5-phenyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD06802815
  • Cas Number : 109496-98-2
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40895
Availability details for
Quantity
  • Availability
  • Add to Cart