ENA183889303 Enamine

3-[4-(1-aminoethyl)phenyl]-1-phenylurea

  • MDL number MFCD09741946
  • Cas Number : 954568-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40919
Availability details for
Quantity
  • Availability
  • Add to Cart