ENA183889304 Enamine

2-amino-4-(difluoromethoxy)-5-methoxybenzoic acid

  • MDL number MFCD08444633
  • Cas Number : 923145-68-0
  • Molecular Weight : 233.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40922
Availability details for
Quantity
  • Availability
  • Add to Cart