ENA183889305 Enamine

4-[4-(azepane-1-sulfonyl)phenyl]-1,3-thiazol-2-amine

  • MDL number MFCD07673083
  • Cas Number : 876706-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40924
Availability details for
Quantity
  • Availability
  • Add to Cart