ENA183889336 Enamine

2-(4-fluorophenyl)-5-(trimethylsilyl)-1,3,4-oxadiazole

  • MDL number MFCD06660928
  • Cas Number : 890405-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37249
Availability details for
Quantity
  • Availability
  • Add to Cart