ENA183889338 Enamine

1-pyrimidin-2-ylpiperidin-4-amine

  • MDL number MFCD06616818
  • Cas Number : 412355-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37313

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37313
Availability details for
Quantity
  • Availability
  • Add to Cart