ENA184121125 Enamine

1-(2-fluoro-5-nitrophenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD11857763
  • Cas Number : 1114822-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33399
Availability details for
Quantity
  • Availability
  • Add to Cart