ENA184121128 Enamine

1-{[2-chloro-5-(trifluoromethyl)phenyl]sulfonyl}piperazine hydrochloride

  • MDL number MFCD11857772
  • Cas Number : 1170080-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40142
Availability details for
Quantity
  • Availability
  • Add to Cart