ENA184121135 Enamine

{4-[(difluoromethyl)sulfanyl]phenyl}methanol

  • MDL number MFCD11857780
  • Cas Number : 1114822-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40955
Availability details for
Quantity
  • Availability
  • Add to Cart