ENA184121136 Enamine

[(4-nitrophenyl)methyl]hydrazine hydrochloride

  • MDL number MFCD11857781
  • Cas Number : 1727-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40971
Availability details for
Quantity
  • Availability
  • Add to Cart