ENA184121138 Enamine

2-amino-2-[4-(trifluoromethyl)phenyl]propanoic acid hydrochloride

  • MDL number MFCD11857782
  • Cas Number : 1171929-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40995
Availability details for
Quantity
  • Availability
  • Add to Cart