ENA184121151 Enamine

2-(3-methyl-1H-1,2,4-triazol-5-yl)aniline

  • MDL number MFCD11839769
  • Cas Number : 25568-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-154
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41152
Availability details for
Quantity
  • Availability
  • Add to Cart