ENA184121158 Enamine

4,6-dimethyl-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile

  • MDL number MFCD11837415
  • Cas Number : 1114822-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41168
Availability details for
Quantity
  • Availability
  • Add to Cart