ENA184121159 Enamine

{4-[(oxolan-2-yl)methoxy]phenyl}methanamine hydrochloride

  • MDL number MFCD11802531
  • Cas Number : 1172099-78-3
  • Molecular Weight : 243.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-41172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41172
Availability details for
Quantity
  • Availability
  • Add to Cart