ENA184121162 Enamine

1-(2-chloroethoxy)-2-methylcyclohexane

  • MDL number MFCD11164766
  • Cas Number : 1094273-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41177
Availability details for
Quantity
  • Availability
  • Add to Cart