ENA184121165 Enamine

methyl [4-(chlorosulfonyl)phenoxy]acetate

  • MDL number MFCD08274832
  • Cas Number : 56077-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41183

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41183
Availability details for
Quantity
  • Availability
  • Add to Cart