ENA184121167 Enamine

2-(2-phenyl-2H-1,2,3-triazol-4-yl)acetonitrile

  • MDL number MFCD11838907
  • Cas Number : 64404-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41191
Availability details for
Quantity
  • Availability
  • Add to Cart