ENA184121169 Enamine

6-methyl-2-(methylsulfanyl)pyrimidine-4-carboxylic acid

  • MDL number MFCD11839775
  • Cas Number : 16490-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-41215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41215
Availability details for
Quantity
  • Availability
  • Add to Cart