ENA184121171 Enamine

N-(4-aminophenyl)-3,4-difluorobenzenesulfonamide hydrochloride

  • MDL number MFCD11857798
  • Cas Number : 1172718-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41232

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41232
Availability details for
Quantity
  • Availability
  • Add to Cart