ENA184121179 Enamine

(3-methylbutan-2-yl)hydrazine dihydrochloride

  • MDL number MFCD11857805
  • Cas Number : 1170075-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41270
Availability details for
Quantity
  • Availability
  • Add to Cart