ENA184121182 Enamine

1-(1-{3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}ethyl)piperazine dihydrochloride

  • MDL number MFCD11839810
  • Cas Number : 1169989-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41305
Availability details for
Quantity
  • Availability
  • Add to Cart