ENA184121185 Enamine

N-[3-(3-aminoprop-1-ynyl)phenyl]methanesulfonamide hydrochloride

  • MDL number MFCD11839777
  • Cas Number : 1171333-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41322

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-41322
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart