ENA184121186 Enamine

methyl 2-(cyclopropylmethoxy)acetate

  • MDL number MFCD10693214
  • Cas Number : 836657-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-41343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41343
Availability details for
Quantity
  • Availability
  • Add to Cart