ENA184121232 Enamine

6-(2,2,2-trifluoroethoxy)pyridin-3-amine

  • MDL number MFCD04111379
  • Cas Number : 72617-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41019
Availability details for
Quantity
  • Availability
  • Add to Cart