ENA184121234 Enamine

1-oxo-1,3-dihydro-2-benzofuran-4-sulfonyl chloride

  • MDL number MFCD11857785
  • Cas Number : 1114822-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41021

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41021
Availability details for
Quantity
  • Availability
  • Add to Cart