ENA184121236 Enamine

N-(3-amino-4-methylphenyl)cyclopropanecarboxamide

  • MDL number MFCD09931958
  • Cas Number : 1016721-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41027

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41027
Availability details for
Quantity
  • Availability
  • Add to Cart