ENA184121237 Enamine

N-(4-bromo-2-methylphenyl)methanesulfonamide

  • MDL number MFCD01213917
  • Cas Number : 330468-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41029
Availability details for
Quantity
  • Availability
  • Add to Cart