ENA184121242 Enamine

4-[2-(diethylamino)ethoxy]-3-methoxybenzonitrile

  • MDL number MFCD09706834
  • Cas Number : 102551-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41035
Availability details for
Quantity
  • Availability
  • Add to Cart