ENA184121244 Enamine

1-[4-(4-methylpiperazine-1-carbonyl)phenyl]methanamine

  • MDL number MFCD08444570
  • Cas Number : 923242-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41038
Availability details for
Quantity
  • Availability
  • Add to Cart