ENA184121251 Enamine

methyl 2-[(3-aminophenyl)formamido]acetate

  • MDL number MFCD09048615
  • Cas Number : 56445-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41065
Availability details for
Quantity
  • Availability
  • Add to Cart