ENA184121266 Enamine

piperidine-1-carboxamide

  • MDL number MFCD00075305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06077
Availability details for
Quantity
  • Availability
  • Add to Cart