ENA184121267 Enamine

4-cyano-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD00510133
  • Cas Number : 63877-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41088
Availability details for
Quantity
  • Availability
  • Add to Cart