ENA184121269 Enamine

3-(3-nitrophenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11839756
  • Cas Number : 5198-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41091

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41091
Availability details for
Quantity
  • Availability
  • Add to Cart