ENA184121293 Enamine

5-(2,6-difluorophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD09732902
  • Cas Number : 1052550-71-6
  • Molecular Weight : 226.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-41132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41132
Availability details for
Quantity
  • Availability
  • Add to Cart