ENA184121299 Enamine

1,1-dioxo-1lambda6-thiolane-3-sulfonamide

  • MDL number MFCD00154901
  • Cas Number : 17115-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41139
Availability details for
Quantity
  • Availability
  • Add to Cart