ENA184121308 Enamine

[(aminosulfonyl)(methyl)amino]acetic acid

  • MDL number MFCD09935151
  • Cas Number : 1016775-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39619

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39619
Availability details for
Quantity
  • Availability
  • Add to Cart