ENA184121315 Enamine

5-isopropyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD02224142
  • Cas Number : 92933-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41163

Safety Information

signalword Warning
safety_statement 36/37
msds https://www.enaminestore.com/catalog/EN300-41163
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart