ENA184121319 Enamine

2-(piperidin-4-yl)-5-(trifluoromethyl)-1,3-benzothiazole

  • MDL number MFCD08271868
  • Cas Number : 400797-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41167
Availability details for
Quantity
  • Availability
  • Add to Cart