ENA184121321 Enamine

(acetyloxy)[4-(chlorosulfonyl)phenyl]methyl acetate

  • MDL number MFCD09991564
  • Cas Number : 69232-47-9
  • Molecular Weight : 306.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-113
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41169

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-41169
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart