ENA184121332 Enamine

5-chloro-2-(pyrrolidin-1-ylcarbonyl)aniline

  • MDL number MFCD09048733
  • Cas Number : 898541-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40487

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40487
Availability details for
Quantity
  • Availability
  • Add to Cart