ENA184121334 Enamine

3,6-dimethyl-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD11857794
  • Cas Number : 80195-31-9
  • Molecular Weight : 206.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41184
Availability details for
Quantity
  • Availability
  • Add to Cart