ENA184121337 Enamine

2-[2-(dimethylamino)ethoxy]benzoic acid hydrochloride

  • MDL number MFCD09997650
  • Cas Number : 1177320-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41192

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41192
Availability details for
Quantity
  • Availability
  • Add to Cart